missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-HSP90 alpha, Polyclonal, Novus Biologicals™

$139.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of the human Hsp90A protein (between residues 650-732) [UniProt P07900]
Antigen HSP90 alpha
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP177685SS Novus Biologicals Canada
NBP177685SS
0.025mL Each for $139.00
NBP177685 Novus Biologicals Canada
NBP177685
0.1mL Each for $309.05
Specifications Description & Specifications

Specifications

Rabbit
HSP90 alpha
Affinity Purified
RUO
P07900
PBS, 30% glycerol
A synthetic peptide made to a C-terminal portion of the human Hsp90A protein (between residues 650-732) [UniProt P07900]
Polyclonal
Primary
Immunocytochemistry,Immunofluorescence,Immunohistochemistry,Immunohistochemistry (Paraffin),Simple Western,Western Blot
3320
1.15mg/mL