Please login to your online account to display your discounted pricing

anti-HSP90 alpha, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an N-terminal portion of the human Hsp90A protein (between residues 1-70) [UniProt P07900]
Antigen HSP90 alpha
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP177682

 
novus biologicals canada
NBP177682
Unlabeled 0.1mL Each for $309.05

NBP177682SS

 
novus biologicals canada
NBP177682SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an N-terminal portion of the human Hsp90A protein (between residues 1-70) [UniProt P07900]
Antigen HSP90 alpha
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Gene Accession No. P07900
Gene ID (Entrez) 3320
Formulation PBS, 30% glycerol
Concentration 1.14mg/mL