Please login to your online account to display your discounted pricing

anti-HSP90 alpha, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP177685SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP177685SS

  • / Each
 

Description & Specifications

Specifications

Antigen HSP90 alpha
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blot
Concentration 1.15mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Format Affinity Purified
Formulation PBS, 30% glycerol
Gene Accession No. P07900
Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of the human Hsp90A protein (between residues 650-732) [UniProt P07900]
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 3320
Target Species Human
Target Species Murine
Target Species Primate
Target Species Rat