Please login to your online account to display your discounted pricing

anti-HSP70/HSPA1A, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminus portion of the human Hsp70 protein (between residues 600-641) [UniProt P08107]
Antigen HSP70/HSPA1A
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP177456SS

 
novus biologicals canada
NBP177456SS
Unlabeled 0.025mL Each for $109.00

NBP177456

 
novus biologicals canada
NBP177456
Unlabeled 0.1mL Each for $309.05
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminus portion of the human Hsp70 protein (between residues 600-641) [UniProt P08107]
Antigen HSP70/HSPA1A
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blotting
Gene Accession No. P08107
Gene ID (Entrez) 3303
Formulation PBS, 30% glycerol
Concentration 1.01mg/mL