Please login to your online account to display your discounted pricing

anti-HSP70/HSPA1A, Clone: 242707, R&D Systems


Manufacturer: r & d systems  RDSMAB1663

 View more versions of this product

Catalog No. MAB1663

  • / Each
 

Description & Specifications

Specifications

Antigen HSP70/HSPA1A
Format Purified
Host Species Mouse
Isotype IgG2a
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Immunogen E.Coli-derived recombinant human HSP70/HSPA1A. Met1-Asp641
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100μg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone 242707

HSP70/HSPA1A Monoclonal antibody MAB1663 Specifically detects HSP70/HSPA1A in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blotting