Please login to your online account to display your discounted pricing

anti-HSP70/HSPA1A, Clone: 242707, R&D Systems

$202.89 - $343.25

Specifications

Antigen HSP70/HSPA1A
Clone 242707
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

MAB1663

 
r & d systems
RDSMAB1663
Unlabeled 100μg Each for $343.25

A400

 
r & d systems
RDSA400
Unlabeled 50μg Each for $202.89
Description & Specifications

Specifications

Antigen HSP70/HSPA1A
Clone 242707
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
Host Species Mouse
Regulatory Status RUO
Immunogen Met1-Asp641
Isotype IgG2a
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Purification Method Protein A or G purified from hybridoma culture supernatant

Applications:

Immunohistochemistry, Western Blotting