Please login to your online account to display your discounted pricing

anti-HSP60, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP177396SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP177396SS

  • / Each
 

Description & Specifications

Specifications

Antigen HSP60
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blot
Concentration 1.0mg/mL
Conjugate Unlabeled
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Format Affinity Purified
Formulation PBS, 30% glycerol
Gene Accession No. P10809
Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of the human Hsp60 protein (within residues 300-360). [Swiss-Prot P10809]
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 3329
Test Specificity Human, mouse, rat, hamster, and primate. Predicted to react with bovine based on 100% sequence homology.
Target Species Hamster
Target Species Human
Target Species Murine
Target Species Primate
Target Species Rat