Please login to your online account to display your discounted pricing

anti-HSP60, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056470SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056470T

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Concentration 0.5mg/mL
Primary or Secondary Primary
Gene Accession No. P10809
Gene ID (Entrez) 3329
Formulation PBS containing 0.2% gelatin
Immunogen A portion of amino acids 150-200 of human Hsp60 was used as the immunogen.
Test Specificity The amino acid sequence used as immunogen is 100% homologous in a broad range of species including human, dog, mouse, rat, bovine, chimp, monkey, and orangutan. Cross reacts with Bovine, Broad, Chimpanzee, Dog, Human and New World Monkey. Predicted to react with Chicken.
Antigen HSP60
Regulatory Status RUO
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Primate
Format Purified
Quantity 0.025mg
Monoclonal or Polyclonal Polyclonal
Applications Western Blotting
Host Species Rabbit