Please login to your online account to display your discounted pricing

anti-HSP60, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056470

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056470

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. P10809
Conjugate Unlabeled
Formulation PBS containing 0.2% gelatin
Gene ID (Entrez) 3329
Test Specificity The amino acid sequence used as immunogen is 100% homologous in a broad range of species including human, dog, mouse, rat, bovine, chimp, monkey, and orangutan. Cross reacts with Bovine, Broad, Chimpanzee, Dog, Human and New World Monkey. Predicted to react with Chicken.
Immunogen A portion of amino acids 150-200 of human Hsp60 was used as the immunogen.
Concentration 0.5mg/mL
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Primate
Host Species Rabbit
Regulatory Status RUO
Antigen HSP60
Format Purified
Applications Western Blotting
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Quantity 0.1mg