Please login to your online account to display your discounted pricing

anti-HSP60, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056469SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056469T

  • / Each
 

Description & Specifications

Specifications

Antigen HSP60
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Primate
Format Purified
Formulation PBS containing 0.2% gelatin
Gene Accession No. P10809
Host Species Rabbit
Immunogen A portion of amino acids 350-400 of human Hsp60 was used as the immunogen.
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 3329
Test Specificity The amino acid sequence used as immunogen is 100% homologous in a broad range of species including human, dog, mouse, rat, bovine, chicken, chimp, monkey, and orangutan. Cross reacts with Bovine, Chicken, Chimpanzee, Dog, Human and New Word Monkey.
Target Species Bovine
Target Species Canine
Target Species Chicken
Target Species Human
Target Species Primate