Please login to your online account to display your discounted pricing

anti-HSP60, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 150-200 of human Hsp60 was used as the immunogen.
Antigen HSP60
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056470T

 
novus biologicals canada
NB10056470SS
Unlabeled 0.025mg Each for $139.00

NB10056470

 
novus biologicals canada
NB10056470
Unlabeled 0.1mg Each for $255.80
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 150-200 of human Hsp60 was used as the immunogen.
Antigen HSP60
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in a broad range of species including human, dog, mouse, rat, bovine, chimp, monkey, and orangutan. Cross reacts with Bovine, Broad, Chimpanzee, Dog, Human and New World Monkey. Predicted to react with Chicken.
Gene Accession No. P10809
Gene ID (Entrez) 3329
Formulation PBS containing 0.2% gelatin
Concentration 0.5mg/mL