missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-HSP60, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 350-400 of human Hsp60 was used as the immunogen.
Antigen HSP60
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB10056469 Novus Biologicals Canada
NB10056469
0.1mg Each for $255.80
NB10056469T Novus Biologicals Canada
NB10056469SS
0.025mg Each for $139.00
Specifications Description & Specifications

Specifications

Rabbit
HSP60
Purified
RUO
The amino acid sequence used as immunogen is 100% homologous in a broad range of species including human, dog, mouse, rat, bovine, chicken, chimp, monkey, and orangutan. Cross reacts with Bovine, Chicken, Chimpanzee, Dog, Human and New Word Monkey.
3329
0.5mg/mL
A portion of amino acids 350-400 of human Hsp60 was used as the immunogen.
Polyclonal
Primary
Western Blot
P10809
PBS containing 0.2% gelatin