Please login to your online account to display your discounted pricing

anti-HSP27, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224533SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224533SS

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Gene ID (Entrez) 3315
Monoclonal or Polyclonal Polyclonal
Cross Reactivity Canine
Cross Reactivity Equine
Cross Reactivity Human
Cross Reactivity Primate
Concentration 0.5mg/mL
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Gene Accession No. P04792
Primary or Secondary Primary
Test Specificity The amino acid sequence used as immunogen for the Neuropilin 1 antibody is 100% homologous in mouse, rat and 92% homologous in human.
Regulatory Status RUO
Immunogen A portion of amino acids 50-100 of human HSP27 was used as the immunogen for the antibody.
Host Species Rabbit
Quantity 0.025mg
Format Affinity Purified
Antigen HSP27