Please login to your online account to display your discounted pricing

anti-HSP27, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224533

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224533

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Concentration 0.5mg/mL
Gene Accession No. P04792
Test Specificity The amino acid sequence used as immunogen for the Neuropilin 1 antibody is 100% homologous in mouse, rat and 92% homologous in human.
Conjugate Unlabeled
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Gene ID (Entrez) 3315
Host Species Rabbit
Cross Reactivity Canine
Cross Reactivity Equine
Cross Reactivity Human
Cross Reactivity Primate
Quantity 0.1mg
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Regulatory Status RUO
Immunogen A portion of amino acids 50-100 of human HSP27 was used as the immunogen for the antibody.
Antigen HSP27