missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-HSP27, Polyclonal, Novus Biologicals™

$139.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of the human Hsp27 protein (within residues 80-150). [Swiss-Prot P04792]
Antigen HSP27
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP175477SS Novus Biologicals Canada
NBP175477SS
0.025mL Each for $139.00
NBP175477 Novus Biologicals Canada
NBP175477
0.1mL Each for $309.05
Specifications Description & Specifications

Specifications

Rabbit
HSP27
Affinity Purified
RUO
Human, mouse and rat. Immunogen has 94% identity to canine and hamster.
3315
1mg/mL
A synthetic peptide made to an internal region of the human Hsp27 protein (within residues 80-150). [Swiss-Prot P04792]
Polyclonal
Primary
Immunocytochemistry,Immunofluorescence,Immunohistochemistry,Immunohistochemistry (Paraffin),Western Blot
P04792
PBS, 30% glycerol