Please login to your online account to display your discounted pricing

anti-HSP27, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 50-100 of human HSP27 was used as the immunogen for the antibody.
Antigen HSP27
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224533SS

 
novus biologicals canada
NBP224533SS
Unlabeled 0.025mg Each for $109.00

NBP224533

 
novus biologicals canada
NBP224533
Unlabeled 0.1mg Each for $257.41
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 50-100 of human HSP27 was used as the immunogen for the antibody.
Antigen HSP27
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen for the Neuropilin 1 antibody is 100% homologous in mouse, rat and 92% homologous in human.
Gene Accession No. P04792
Gene ID (Entrez) 3315
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL