Please login to your online account to display your discounted pricing

anti-HRH1/H1 Receptor Polyclonal, Thermo Scientific™

HRH1/H1 Receptor Polyclonal Ab PA5-33842 from Thermo Scientific™ specifically detects HRH1/H1 Receptor in Hu samples, and it is validated for IHC(P)

$506.05 - $506.05

Specifications

Antigen HRH1/H1 Receptor
Cross Reactivity Human
Format Purified
Host Species Rabbit
Immunogen Synthetic 19 amino acid peptide from 3rd cytoplasmic domain of human HRH1/Histamine H1 Receptor.
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

PIPA533842

 
pierce biotechnology inc
PA533842
Unlabeled 50μg Each for $506.05

PIPA533843

 
pierce biotechnology inc
PA533843
Unlabeled 50μg Each for $506.05
Description & Specifications

Specifications

Antigen HRH1/H1 Receptor
Cross Reactivity Human
Format Purified
Host Species Rabbit
Immunogen Synthetic 19 amino acid peptide from 3rd cytoplasmic domain of human HRH1/Histamine H1 Receptor.
Isotype IgG
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements Store at 2-8°C for up to 1 month. For long term storage store at -20°C

HRH1/H1 Receptor Polyclonal Ab PA5-33842 from Thermo Scientific™ specifically detects HRH1/H1 Receptor in Hu samples, and it is validated for IHC(P)

Pheromones are chemical signals that elicit specific behavioral responses and physiologic alterations in recipients of the same species. The protein encoded by this gene is similar to pheromone receptors and is primarily localized to the olfactory mucosa. An alternate splice variant of this gene is thought to exist, but its full length nature has not been determined. Putative pheromone receptor. Expressed in the olfactory mucosa, very low expression in brain, lung and kidney. The chimpanzee and orangutan orthologous proteins do not exist, their genes are pseudogenes.
Host Species: Rabbit
Cross Reactivity: Human
Isotype: IgG
Immunogen: Synthetic 17 amino acid peptide from 2nd extracellular domain of human HRH1/Histamine H1 Receptor.
Formulation: 50μg purified Antibody (1mg/mL) in PBS, with < 0.1% sodium azide;
Synonyms: HRH1, H1 histamine receptor, H1 receptor, Histamine 1 receptor, Histamine H1 receptor, H1-R, H1R, Histamine H(1) receptor, Histamine receptor H1, HisH1
Uniprot Accession: P35367
Gene Symbol: HRH1
Entrez GeneID: 3269
PA5-34308 detects V1RL1/VN1R1 in Human samples. PA5-34308 has been successfully used in Immunohistochemistry (Paraffin) procedures. PA5-34308 The immunogen is Synthetic 17 amino acid peptide from 2nd extracellular domain of human VN1R1..