missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-HRD1, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of human HRD1 (within residues 300-400). [UniProt Q96PK3]
Antigen HRD1
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB1002526SS Novus Biologicals Canada
NB1002526SS
0.025mL Each for $139.00
NB1002526 Novus Biologicals Canada
NB1002526
0.1mL Each for $329.00
Specifications Description & Specifications

Specifications

Rabbit
HRD1
Antisera
RUO
Q86TM6
A synthetic peptide made to an internal portion of human HRD1 (within residues 300-400). [UniProt Q96PK3]
Polyclonal
Primary
Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Paraffin),Western Blot
84447