Please login to your online account to display your discounted pricing

anti-HP1 alpha, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224601

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224601

  • / Each
 

Description & Specifications

Specifications

Antigen HP1 alpha
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Format Purified
Formulation 100μg in 200μL PBS containing 0.05% BSA
Gene Accession No. P45973
Host Species Rabbit
Immunogen A portion of amino acids 150-200 of human CBX5 was used as the immunogen.
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 23468
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, monkey, chimpanzee and chicken, and 94% homologous in horse, cow, mouse and rat.
Target Species Canine
Target Species Human
Target Species Murine
Target Species Primate