Please login to your online account to display your discounted pricing

anti-HP1 alpha, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 50-100 of human CBX5 was used as the immunogen.
Antigen HP1 alpha
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224602

 
novus biologicals canada
NBP224602
Unlabeled 0.1mg Each for $257.41

NBP224602SS

 
novus biologicals canada
NBP224602SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 50-100 of human CBX5 was used as the immunogen.
Antigen HP1 alpha
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_a, CRA_b and CRA_c), chimpanzee and monkey, 94% homologous in cow and 83% homologous in mouse and rat.
Gene Accession No. P45973
Gene ID (Entrez) 23468
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL