missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-HP1 alpha, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 150-200 of human CBX5 was used as the immunogen.
Antigen HP1 alpha
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP224601 Novus Biologicals Canada
NBP224601
0.1mg Each for $257.41
NBP224601SS Novus Biologicals Canada
NBP224601SS
0.025mg Each for $109.00
Specifications Description & Specifications

Specifications

Rabbit
HP1 alpha
Purified
RUO
The amino acid sequence used as immunogen is 100% homologous in human, monkey, chimpanzee and chicken, and 94% homologous in horse, cow, mouse and rat.
23468
0.5mg/mL
A portion of amino acids 150-200 of human CBX5 was used as the immunogen.
Polyclonal
Primary
Western Blot
P45973
100μg in 200μL PBS containing 0.05% BSA