Please login to your online account to display your discounted pricing

anti-HP1 alpha, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide containing an N-terminal region of human HP1 alpha (within residues 1-100). [Swiss-Prot P45973]
Antigen HP1 alpha
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB1002518

 
novus biologicals canada
NB1002518
Unlabeled 0.1mL Each for $329.00

NB1002518SS

 
novus biologicals canada
NB1002518SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide containing an N-terminal region of human HP1 alpha (within residues 1-100). [Swiss-Prot P45973]
Antigen HP1 alpha
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Test Specificity Human. Expected to react with mouse, Arabidopsis suecica, Rye and Triticum aestivum due to sequence homology.
Gene Accession No. P45973
Gene ID (Entrez) 23468
Formulation PBS