missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-HOXD8, Polyclonal, Novus Biologicals™

$139.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 250-328 of human HOXD8 was used as the immunogen. 264 281
Antigen HOXD8
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP227108 Novus Biologicals Canada
NBP227108
0.1mg Each for $257.41
NBP227108SS Novus Biologicals Canada
NBP227108SS
0.025mg Each for $139.00
Specifications Description & Specifications

Specifications

Rabbit
HOXD8
Purified
RUO
The amino acid sequence used as immunogen is 100% homologous in human and chimpanzee, 94% homologous in cow, 74% homologous in opossum and 66% in mouse.
3234
0.5mg/mL
A portion of amino acids 250-328 of human HOXD8 was used as the immunogen. 264 281
Polyclonal
Primary
Western Blot
P13378
100μg in 200μL PBS containing 0.05% BSA.