Please login to your online account to display your discounted pricing

anti-HOXD13, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224889SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224889SS

  • / Each
 

Description & Specifications

Specifications

Antigen HOXD13
Applications Chromatin Immunoprecipitation
Applications Immunoprecipitation
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Format Affinity Purified
Formulation 0.1mg antibody in 0.2mL PBS containing 0.2% gelatin
Gene Accession No. P35453
Host Species Rabbit
Immunogen This antibody was developed against a synthetic peptide corresponding to amino acids 184-207 of human HOXD13; Genbank accession no. NP_000514.1.
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 3239
Test Specificity This peptide sequence is reported by NCBI to be 100% conserved between human, chimpanzee, rat and mouse HOXD13.