Please login to your online account to display your discounted pricing

anti-HOXD13, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224889

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224889

  • / Each
 

Description & Specifications

Specifications

Formulation 0.1mg antibody in 0.2mL PBS containing 0.2% gelatin
Gene Accession No. P35453
Host Species Rabbit
Concentration 0.5mg/mL
Antigen HOXD13
Primary or Secondary Primary
Test Specificity This peptide sequence is reported by NCBI to be 100% conserved between human, chimpanzee, rat and mouse HOXD13.
Gene ID (Entrez) 3239
Conjugate Unlabeled
Quantity 0.1mg
Applications Chromatin Immunoprecipitation
Applications Immunoprecipitation
Immunogen This antibody was developed against a synthetic peptide corresponding to amino acids 184-207 of human HOXD13; Genbank accession no. NP_000514.1.
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Regulatory Status RUO
Format Affinity Purified
Monoclonal or Polyclonal Polyclonal