Please login to your online account to display your discounted pricing

anti-HOXD13, Polyclonal, Novus Biologicals™

$139.00 - $321.00

Specifications

Host Species Rabbit
Immunogen This antibody was developed against a synthetic peptide corresponding to amino acids 184-207 of human HOXD13; Genbank accession no. NP_000514.1.
Antigen HOXD13
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224889SS

 
novus biologicals canada
NBP224889SS
Unlabeled 0.025mg Each for $139.00

NBP224889

 
novus biologicals canada
NBP224889
Unlabeled 0.1mg Each for $321.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen This antibody was developed against a synthetic peptide corresponding to amino acids 184-207 of human HOXD13; Genbank accession no. NP_000514.1.
Antigen HOXD13
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Chromatin Immunoprecipitation
Applications Immunoprecipitation
Test Specificity This peptide sequence is reported by NCBI to be 100% conserved between human, chimpanzee, rat and mouse HOXD13.
Gene Accession No. P35453
Gene ID (Entrez) 3239
Formulation 0.1mg antibody in 0.2mL PBS containing 0.2% gelatin
Concentration 0.5mg/mL