Please login to your online account to display your discounted pricing

anti-HOXB4, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224561SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224561SS

  • / Each
 

Description & Specifications

Specifications

Formulation 100μg in 200μL PBS containing 0.05% BSA
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, mouse, rat, chimp, cat, dogand horse.
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 3214
Immunogen A portion of amino acids 100-150 of human HOXB4 was used as the immunogen.
Concentration 0.5mg/mL
Cross Reactivity Bovine
Cross Reactivity Feline
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Conjugate Unlabeled
Gene Accession No. P17483
Regulatory Status RUO
Format Affinity Purified
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Antigen HOXB4
Primary or Secondary Primary
Host Species Rabbit
Quantity 0.025mg