missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-HOXB4, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 1-50 of human HOXB4 was used as the immunogen.
Antigen HOXB4
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP224598 Novus Biologicals Canada
NBP224598
0.1mg Each for $257.41
NBP224598SS Novus Biologicals Canada
NBP224598SS
0.025mg Each for $109.00
Specifications Description & Specifications

Specifications

Rabbit
HOXB4
Purified
RUO
The amino acid sequence used as immunogen is 100% homologous in human, rhesus monkey and chimpanzee, 94% homologous in cow, rat, mouse and wild boar, 88% homologous in horse and 76% homologous in opossum.
3214
0.5mg/mL
A portion of amino acids 1-50 of human HOXB4 was used as the immunogen.
Polyclonal
Primary
Immunohistochemistry (Paraffin)
P17483
100μg in 200μL PBS containing 0.05% BSA