Please login to your online account to display your discounted pricing

anti-HOXB4, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 100-150 of human HOXB4 was used as the immunogen.
Antigen HOXB4
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224561SS

 
novus biologicals canada
NBP224561SS
Unlabeled 0.025mg Each for $109.00

NBP224561

 
novus biologicals canada
NBP224561
Unlabeled 0.1mg Each for $255.80
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 100-150 of human HOXB4 was used as the immunogen.
Antigen HOXB4
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, mouse, rat, chimp, cat, dogand horse.
Gene Accession No. P17483
Gene ID (Entrez) 3214
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL