Please login to your online account to display your discounted pricing

anti-HOXB4, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224598SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224598SS

  • / Each
 

Description & Specifications

Specifications

Antigen HOXB4
Applications Immunohistochemistry (Paraffin)
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Zebrafish
Format Purified
Formulation 100μg in 200μL PBS containing 0.05% BSA
Gene Accession No. P17483
Host Species Rabbit
Immunogen A portion of amino acids 1-50 of human HOXB4 was used as the immunogen.
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 3214
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, rhesus monkey and chimpanzee, 94% homologous in cow, rat, mouse and wild boar, 88% homologous in horse and 76% homologous in opossum.
Target Species Canine
Target Species Human
Target Species Murine
Target Species Rat
Target Species Zebrafish