Please login to your online account to display your discounted pricing

anti-HOXB4, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224598

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224598

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Gene ID (Entrez) 3214
Concentration 0.5mg/mL
Formulation 100μg in 200μL PBS containing 0.05% BSA
Conjugate Unlabeled
Quantity 0.1mg
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Zebrafish
Gene Accession No. P17483
Host Species Rabbit
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, rhesus monkey and chimpanzee, 94% homologous in cow, rat, mouse and wild boar, 88% homologous in horse and 76% homologous in opossum.
Monoclonal or Polyclonal Polyclonal
Immunogen A portion of amino acids 1-50 of human HOXB4 was used as the immunogen.
Applications Immunohistochemistry (Paraffin)
Format Purified
Regulatory Status RUO
Antigen HOXB4