Please login to your online account to display your discounted pricing

anti-HOXB13, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB1002492SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB1002492SS

  • / Each
 

Description & Specifications

Specifications

Formulation Rabbit sera with 50% glycerol
Gene Accession No. NP_006352
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 10481
Regulatory Status RUO
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Test Specificity Reacts with Human. The amino acid sequence used as immunogen is 100% homologous in human, mouse, and rat, and 94% homologous in zebrafish, pufferfish, medaka, bovine and dog for HOXB13. It is 88% homologous in human, mouse, rat, dog, chicken, chimp, bovine and zebrafish for HOXA13. It is 77% homologous in human and mouse for HOXC13.
Conjugate Unlabeled
Antigen HOXB13
Format Antiserum
Quantity 0.025mL
Applications ELISA
Applications Western Blotting
Primary or Secondary Primary
Host Species Rabbit
Immunogen Amino acids 220-260 of human HOXB13 protein were used as the immunogen.