Please login to your online account to display your discounted pricing

anti-HOXB13, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB1002492

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB1002492

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. NP_006352
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Formulation Rabbit sera with 50% glycerol
Host Species Rabbit
Applications ELISA
Applications Western Blotting
Primary or Secondary Primary
Gene ID (Entrez) 10481
Test Specificity Reacts with Human. The amino acid sequence used as immunogen is 100% homologous in human, mouse, and rat, and 94% homologous in zebrafish, pufferfish, medaka, bovine and dog for HOXB13. It is 88% homologous in human, mouse, rat, dog, chicken, chimp, bovine and zebrafish for HOXA13. It is 77% homologous in human and mouse for HOXC13.
Immunogen Amino acids 220-260 of human HOXB13 protein were used as the immunogen.
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Antiserum
Quantity 0.1mL
Regulatory Status RUO
Antigen HOXB13