missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-HOXB13, Polyclonal, Novus Biologicals™

$139.00 - $274.59

Specifications

Host Species Rabbit
Immunogen Amino acids 220-260 of human HOXB13 protein were used as the immunogen.
Antigen HOXB13
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB1002492 Novus Biologicals Canada
NB1002492
0.1mL Each for $274.59
NB1002492SS Novus Biologicals Canada
NB1002492SS
0.025mL Each for $139.00
Specifications Description & Specifications

Specifications

Rabbit
HOXB13
Antisera
RUO
Reacts with Human. The amino acid sequence used as immunogen is 100% homologous in human, mouse, and rat, and 94% homologous in zebrafish, pufferfish, medaka, bovine and dog for HOXB13. It is 88% homologous in human, mouse, rat, dog, chicken, chimp, bovine and zebrafish for HOXA13. It is 77% homologous in human and mouse for HOXC13.
10481
Amino acids 220-260 of human HOXB13 protein were used as the immunogen.
Polyclonal
Primary
ELISA,Western Blot
NP_006352
Rabbit sera with 50% glycerol