Please login to your online account to display your discounted pricing

anti-HOXA9, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224633

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224633

  • / Each
 

Description & Specifications

Specifications

Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, chimp, rhesus monkey, cat and horse, 92% homologous in dog and 66% homologous in mouse.
Concentration 0.5mg/mL
Host Species Rabbit
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene Accession No. P31269
Formulation 100μg in 200μL PBS containing 0.05% BSA
Conjugate Unlabeled
Gene ID (Entrez) 3205
Format Purified
Immunogen A portion of amino acids 225-272 of human HOXA9 was used as the immunogen.
Cross Reactivity Chicken
Cross Reactivity Feline
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Applications Western Blotting
Regulatory Status RUO
Antigen HOXA9
Quantity 0.1mg