missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-HOXA9, Polyclonal, Novus Biologicals™

$139.00 - $259.02

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 180-230 of human HOXA9 was used as the immunogen for this antibody.
Antigen HOXA9
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP224663 Novus Biologicals Canada
NBP224663
0.1mg Each for $259.02
NBP224663SS Novus Biologicals Canada
NBP224663SS
0.025mg Each for $139.00
Specifications Description & Specifications

Specifications

Rabbit
HOXA9
Purified
RUO
The amino acid sequence used as immunogen for this antibody is 100% homologous in variola virus, 93% homologus incowpox, camelpox and taderapox virus,86% homologus invaccinia virus and 80% homologus in ectromelia, rabbitpox and monkeypox virus.
3205
0.5mg/mL
A portion of amino acids 180-230 of human HOXA9 was used as the immunogen for this antibody.
Polyclonal
Primary
Immunohistochemistry (Paraffin)
P31269
100μg antibody in 200μL PBS containing 0.05% BSA