Please login to your online account to display your discounted pricing

anti-HOXA9, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 225-272 of human HOXA9 was used as the immunogen.
Antigen HOXA9
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224633

 
novus biologicals canada
NBP224633
Unlabeled 0.1mg Each for $255.80

NBP224633SS

 
novus biologicals canada
NBP224633SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 225-272 of human HOXA9 was used as the immunogen.
Antigen HOXA9
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, chimp, rhesus monkey, cat and horse, 92% homologous in dog and 66% homologous in mouse.
Gene Accession No. P31269
Gene ID (Entrez) 3205
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL