Please login to your online account to display your discounted pricing

anti-HOXA9, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224663SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224663SS

  • / Each
 

Description & Specifications

Specifications

Antigen HOXA9
Applications Immunohistochemistry (Paraffin)
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Canine
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Format Purified
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Gene Accession No. P31269
Host Species Rabbit
Immunogen A portion of amino acids 180-230 of human HOXA9 was used as the immunogen for this antibody.
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 3205
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in variola virus, 93% homologus incowpox, camelpox and taderapox virus,86% homologus invaccinia virus and 80% homologus in ectromelia, rabbitpox and monkeypox virus.
Target Species Canine
Target Species Chicken
Target Species Human
Target Species Murine
Target Species Primate
Target Species Rat