Please login to your online account to display your discounted pricing

anti-HOX11, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A peptide sequence corresponding to acids amino acids 306-318 (LQNLQPWSDDSTK) of human HOX11 was used as the immunogen.
Antigen HOX11
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056424

 
novus biologicals canada
NB10056424
Unlabeled 0.1mg Each for $255.80

NB10056424T

 
novus biologicals canada
NB10056424SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A peptide sequence corresponding to acids amino acids 306-318 (LQNLQPWSDDSTK) of human HOX11 was used as the immunogen.
Antigen HOX11
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity Cross reacts with Mouse, Human and Rat. Predicted to react with Bovine, Chicken, Chimpanzee, Dog and Xenopus.
Gene ID (Entrez) 3195
Formulation PBS containing 0.2% gelatin
Concentration 0.5mg/mL