Please login to your online account to display your discounted pricing

anti-HO-1/HMOX1/HSP32, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP177459SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP177459SS

  • / Each
 

Description & Specifications

Specifications

Antigen HO-1/HMOX1/HSP32
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Concentration 1.16mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Affinity Purified
Formulation PBS, 30% glycerol
Gene Accession No. P09601
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human Heme Oxygenase 1 protein (between residues 100-150) [UniProt P09601]
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 3162
Target Species Human
Target Species Murine