Please login to your online account to display your discounted pricing

anti-hnRNP K, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224559SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224559SS

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Immunogen A portion of amino acid 380 to 430 of human hnRNPK was used as immunogen.
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Opossum
Cross Reactivity Primate
Cross Reactivity Rat
Primary or Secondary Primary
Formulation 100μg in 200μL PBS containing 0.05% BSA
Gene ID (Entrez) 3190
Concentration 0.5mg/mL
Isotype IgG
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, mouse, monkey, dog, chimpanzee, rat and cow, 90% homologous in opossum and zebrafish.
Regulatory Status RUO
Quantity 0.025mg
Applications Immunohistochemistry (Paraffin)
Format Affinity Purified
Host Species Rabbit
Monoclonal or Polyclonal Polyclonal
Antigen hnRNP K