Please login to your online account to display your discounted pricing

anti-hnRNP K, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224559

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224559

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Formulation 100μg in 200μL PBS containing 0.05% BSA
Host Species Rabbit
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 3190
Concentration 0.5mg/mL
Primary or Secondary Primary
Quantity 0.1mg
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, mouse, monkey, dog, chimpanzee, rat and cow, 90% homologous in opossum and zebrafish.
Format Affinity Purified
Immunogen A portion of amino acid 380 to 430 of human hnRNPK was used as immunogen.
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Opossum
Cross Reactivity Primate
Cross Reactivity Rat
Applications Immunohistochemistry (Paraffin)
Isotype IgG
Regulatory Status RUO
Antigen hnRNP K