Please login to your online account to display your discounted pricing

anti-hnRNP K, Polyclonal, Novus Biologicals™

$139.00 - $257.41

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A portion of amino acid 380 to 430 of human hnRNPK was used as immunogen.
Antigen hnRNP K
Monoclonal or Polyclonal Polyclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224559

 
novus biologicals canada
NBP224559
Unlabeled 0.1mg Each for $257.41

NBP224559SS

 
novus biologicals canada
NBP224559SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A portion of amino acid 380 to 430 of human hnRNPK was used as immunogen.
Antigen hnRNP K
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, mouse, monkey, dog, chimpanzee, rat and cow, 90% homologous in opossum and zebrafish.
Gene ID (Entrez) 3190
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL