Please login to your online account to display your discounted pricing

anti-hnRNP K, Polyclonal, Novus Biologicals™

$139.00 - $257.41

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A portion of amino acid 200 to 250 of human hnRNPK was used as immunogen for this antibody.
Antigen hnRNP K
Monoclonal or Polyclonal Polyclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224532SS

 
novus biologicals canada
NBP224532SS
Unlabeled 0.025mg Each for $139.00

NBP224532

 
novus biologicals canada
NBP224532
Unlabeled 0.1mg Each for $257.41
Description & Specifications

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A portion of amino acid 200 to 250 of human hnRNPK was used as immunogen for this antibody.
Antigen hnRNP K
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen for the MRF2 (ARID5B) antibody is 100% homologous in human (isoforms CRA_a, CRA_c), chimpanzee and monkey, 93% homologous in mouse and horse and 87% homologous in rat.
Gene ID (Entrez) 3190
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL