Please login to your online account to display your discounted pricing

anti-HMGB3/HMG4, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 150-200 of human HMGB3 was used as the immunogen.
Antigen HMGB3/HMG4
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224512

 
novus biologicals canada
NBP224512
Unlabeled 0.1mg Each for $257.41

NBP224512SS

 
novus biologicals canada
NBP224512SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 150-200 of human HMGB3 was used as the immunogen.
Antigen HMGB3/HMG4
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_c).
Gene Accession No. O15347
Gene ID (Entrez) 3149
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL