missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-HMGB1/HMG-1, PE, Clone: 115603, R&D Systems

$378.79 - $378.79

Specifications

Antigen HMGB1/HMG-1
Clone 115603
Format Purified
Host Species Murine
Regulatory Status RUO
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
IC1690P R & D Systems
RDSIC1690P
100 Tests Each for $378.79
Specifications

Description

Applications:

Chromatin Immunoprecipitation, Flow Cytometry, Immunocytochemistry, Western Blotting

Description & Specifications

Specifications

HMGB1/HMG-1
Purified
RUO
IgG2b
Primary
115603
Murine
E.Coli-derived recombinant human HMGB1/HMG-1. Gly2-Glu215 (predicted)
Monoclonal
Protein A or G purified from hybridoma culture supernatant