Please login to your online account to display your discounted pricing

anti-HMGA1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056468SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056468T

  • / Each
 

Description & Specifications

Specifications

Antigen HMGA1
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rat
Format Affinity Purified
Formulation PBS containing 0.05% BSA
Gene Accession No. NP_665908
Host Species Rabbit
Immunogen A synthetic peptide (DGTEKRGRGRPRKQ) corresponding to amino acids 19-32 of human HMGA1 was used as immunogen.
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 3159
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, dog, mouse, rat, pig, 92% homologous in bovine and 84% homologous in chicken. Reacts with Dog, Human, Mouse, Pig and Rat. Predicted to react with Bovine and Chicken.
Target Species Canine
Target Species Human
Target Species Murine
Target Species Porcine
Target Species Rat