missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-HMGA1, Polyclonal, Novus Biologicals™

$139.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A synthetic peptide (DGTEKRGRGRPRKQ) corresponding to amino acids 19-32 of human HMGA1 was used as immunogen.
Antigen HMGA1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB10056468T Novus Biologicals Canada
NB10056468SS
0.025mg Each for $139.00
NB10056468 Novus Biologicals Canada
NB10056468
0.1mg Each for $257.41
Specifications Description & Specifications

Specifications

Rabbit
HMGA1
Affinity Purified
RUO
The amino acid sequence used as immunogen is 100% homologous in human, dog, mouse, rat, pig, 92% homologous in bovine and 84% homologous in chicken. Reacts with Dog, Human, Mouse, Pig and Rat. Predicted to react with Bovine and Chicken.
3159
0.5mg/mL
A synthetic peptide (DGTEKRGRGRPRKQ) corresponding to amino acids 19-32 of human HMGA1 was used as immunogen.
Polyclonal
Primary
Immunohistochemistry (Paraffin),Western Blot
NP_665908
PBS containing 0.05% BSA