Please login to your online account to display your discounted pricing

anti-Histone H4 (Trimethyl Lys20), Polyclonal, Novus Biologicals™

$219.00 - $384.00

Specifications

Host Species Rabbit
Immunogen Synthetic trimethylated peptide surrounding Lysine 20 of human Histone H4 [Swiss Prot P62805].
Antigen Histone H4 (Trimethyl Lys20)
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB212090

 
novus biologicals canada
NB212090
Unlabeled 0.05mg Each for $384.00

NB212090SS

 
novus biologicals canada
NB212090SS
Unlabeled 0.025mg Each for $219.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic trimethylated peptide surrounding Lysine 20 of human Histone H4 [Swiss Prot P62805].
Antigen Histone H4 (Trimethyl Lys20)
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Dot Blot
Applications Immunohistochemistry
Applications Simple Western
Applications Western Blot
Test Specificity Human, mouse and rat. Predicted to react with most mammal species.
Gene Accession No. P62805
Gene ID (Entrez) 121504
Formulation PBS, 30% glycerol
Concentration 0.58mg/mL