Please login to your online account to display your discounted pricing

anti-Histone H4 (Monomethyl Lys20), Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB212088SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB212088SS

  • / Each
 

Description & Specifications

Specifications

Antigen Histone H4 (Monomethyl Lys20)
Applications Chromatin Immunoprecipitation
Applications Dot Blot
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Concentration 1.12mg/mL
Conjugate Unlabeled
Cross Reactivity C. elegans
Cross Reactivity Human
Cross Reactivity Mouse
Format Affinity Purified
Formulation PBS, 30% glycerol
Gene Accession No. P62805
Host Species Rabbit
Immunogen Synthetic monomethylated peptide surrounding Lysine 20 of human Histone H4 [Swiss Prot P62805].
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 121504
Test Specificity Human, mouse, and S. cerevisiae. Predicted to react with many species including rat, chicken, Xenopus, Drosophila, and plant based on 100% sequence homology.
Target Species C. elegans
Target Species Human
Target Species Murine